Imprimer
Pump modules, C-601 and C-605
Fournisseur: BUCHI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
054105
BUCH054105EA 0 EUR
BUCH054105
Pump modules, C-601 and C-605
Pompe Pompes HPLC
The three-piston pump module with a pulse-free flow rate from 2,5 to 250 ml/min guarantees fast separation at a maximum working pressure of 10 bar (145 psi) or 50 bar (725 psi) for isocratic or gradient mode. These pump modules are the core components of the Sepacore® chromatography systems. The chemically resistant and biocompatible pumps can be used with a wide range of organic solvents.
  • Plug and play portable unit
  • Programs and settings enables the selection of flow rate, pressure, time and gradient of the solvents
  • Material in contact with solvents are
En savoir plus