Imprimer
Anti-rh BDNF Sheep Polyclonal Antibody
Fournisseur: Biosensis
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
S-015-500
BSENS-015-500EA 312 EUR
BSENS-015-500
Anti-rh BDNF Sheep Polyclonal Antibody
Anticorps
BDNF belongs to the neurotrophin family and promotes the survival of neuronal populations that are all located either in the central nervous system or directly connected to it. It is a major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability. The alterations in BDNFEn savoir plus