Imprimer
Anti-RBFOX3 Rabbit Polyclonal Antibody
Fournisseur: Biosensis
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
R-3771-100
BSENR-3771-100EA 312 EUR
BSENR-3771-100
Anti-RBFOX3 Rabbit Polyclonal Antibody
Anticorps
Rabbit anti-neuronal nuclei antigen/Fox3 (NeuN/Fox3) polyclonal antibody (unconjugated), suitable for WB, IHC, ICC.
  • Human, mouse, rat
Applications: Western Blotting (WB), Immunocytochemistry (ICC) and Immunohistochemistry (IHC). A dilution of 1:500 - 1:1,000 is recommended for WB. A dilution of 1:5,000 - 1:10,000 is recommended for ICC and IHC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.