Imprimer
Anti-Myc Tag Rabbit Polyclonal Antibody
Fournisseur: Biosensis
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
R-1319-100
BSENR-1319-100EA 407 EUR
BSENR-1319-100
Anti-Myc Tag Rabbit Polyclonal Antibody
Anticorps
The Myc tag contains the amino acids Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu (E-Q-K-L-I-S-E-E-D-L) corresponding to amino acids 410-419 of human Myc. This tag is widely used for monitoring expression of recombinant proteins in bacteria, insect and mammalian cells.
This polyclonal anti-Myc-tag antibody detects overexpressed proteins containing the Myc epitope tag. The antibody recognizes the Myc-tag fused to either the amino- or carboxy- termini of targeted proteins in transfected or transformed cells.
Application Information:
ELISA and Western Blotting (WB). Suggested dilutions for ELISAEn savoir plus