Imprimer
Anti-Tnfrsf16 Mouse Monoclonal Antibody [clone: MC192] Saporin
Fournisseur: Biosensis
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
M-1755-25-IT M-1755-50-IT M-1755-100-IT
BSENM-1755-25-ITEA 302 EUR
BSENM-1755-25-IT BSENM-1755-50-IT BSENM-1755-100-IT
Anti-Tnfrsf16 Mouse Monoclonal Antibody [clone: MC192] Saporin
Anticorps
MC192-saporin is an antibody conjugate comprising of the monoclonal antibody MC192 against rat p75NTR, the nerve growth factor receptor, chemically conjugated via a reducible disulfide bridge to the ribosome-inactivating protein saporin, purified from saponaria officinalis.
  • Specific only for rat NGFR
  • No reactivity to human or mouse NGFR has been reported
  • Does not cross react with p75NTR-expressing cells in other species than rat
Unconjugated saporin is incapable of entering the cells due to the apparent lack of ligand. Upon specific binding via MC192En savoir plus