Imprimer

GFP PicoKine; ELISA Kit

Marques : Boster Bio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

EK1440
BSBTEK1440EA 788 EUR
BSBTEK1440
GFP PicoKine; ELISA Kit
Tests Tests ELISA

Sandwich high sensitivity ELISA kit for quantitative detection of GFP.


  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts


Detect GFP GFP with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell lysates. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. GFP tissue specificity: Photocytes.


96wells/kit, with removable strips.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR