Imprimer

ELISA Kit, Human Kininogen-1/KNG1 PicoKine

Marques : Boster Bio
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

EK1154
BSBTEK1154EA 699 EUR
BSBTEK1154
ELISA Kit, Human Kininogen-1/KNG1 PicoKine
Tests Tests ELISA

Sandwich high sensitivity ELISA kit for quantitative detection of human kininogen-1/KNG1.


  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts


Detect human kininogen/KNG1 with <20 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Kininogen/KNG1 tissue specificity: secreted in plasma. T-kinin is detected in malignant ovarian, colon and breast carcinomas, but not in benign tumors.


96wells/kit, with removable strips.

Commander maintenant

SPÉCIFICATIONS

Environmentally Preferable
Primary antibody reactivity Human
Type de test Sandwich
Hôte

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR