Imprimer
Micro burettes, Bang pattern, straight stopcock, Schellbach stripe, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
24265 24269 924265
BRND24265EA 420 EUR
BRND24265 BRND24269 BRND924265
Micro burettes, Bang pattern, straight stopcock, Schellbach stripe, BLAUBRAND®
Burettes
Verre borosilicaté 3.3, classe AS,marquage DE-M.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • With base
Certifications: DIN EN ISO 385
Informations de commande: Supplied with batch certificate or single certificate. On request also available with USP individual certificate or DAkkS calibration certificate.