Imprimer
Bulb pipettes, BLAUBRAND®, two marks, Class AS
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2.29722 2.29721 2.29725 2.29738 2.29732 2.29733 2.29734 2.29735 2.29727 2.29723
BRND2.29722EA 115 EUR
BRND2.29722 BRND2.29721 BRND2.29725 BRND2.29738 BRND2.29732 BRND2.29733 BRND2.29734 BRND2.29735 BRND2.29727 BRND2.29723
Bulb pipettes, BLAUBRAND®, two marks, Class AS
Pipettes Pipettes jaugées
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Supplied with LOT certificate
Certifications: DIN EN ISO 648; DE-M marking