Imprimer
Quick Charger US for M410 Label Printer
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
176994
BRDY176994EA 213 EUR
BRDY176994
Quick Charger US for M410 Label Printer
Piles
This quick charger charges the rechargeable NiMH battery in the M410 label printer faster than the standard AC adaptor.
  • With a quick charger on hand, you can be sure your battery is ready to go when you need it most
  • Plugs directly into battery and charges battery from a wall outlet rather than from inside the printer
This quick charger charges the rechargeable NiMH battery in the M410 Label Printer faster than the standard AC adaptor. Package includes the Quick Charger only (printer and battery sold separately). With a quick charger on hand, you canEn savoir plus