Imprimer
Brady® Water Soluble Paper Label Tapes for BradyJet™ J7300 Colour Label Printer
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
170461 176583 170460 176605 170462
BRDY170461EA 104 EUR
BRDY170461 BRDY176583 BRDY170460 BRDY176605 BRDY170462
Brady® Water Soluble Paper Label Tapes for BradyJet™ J7300 Colour Label Printer
Étiquettes
Choose water soluble paper labels for complete label and adhesive removal. Ideal for laboratories, temporary identification needs, and work-in-process product flows.
  • Label and adhesive completely remove in 30 seconds using warm water
  • Inkjet receptive labels resist abrasions, humidity, and temperatures from −70 to +100 °C
  • Designed for indoor use
Dissolvable acrylic adhesive is ideal for labs, asset tracking, and temporary ID. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, andEn savoir plus