Imprimer
BradyJet J4000-EU Label Printer, Colour
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
152711
BRDY152711EA 3420 EUR
BRDY152711
BradyJet J4000-EU Label Printer, Colour
Imprimantes Label Printers
High-impact, precision full colour that lasts. Create the compliant, long-lasting labels, signs and tags you need with the BradyJet J4000 inkjet printer.
  • Full colour
  • Long-lasting
  • 5000 labels per day
This industry inkjet workhorse creates up to 5000 impactful full-colour labels per day up to 203,20 mm wide on continuous or die-cut material. Ideal for a wide variety of applications in safety, including lean/5S, lockout tagout, instructional or safety chemical labelling for handling, storage or transport, as well as lab settings. Contains the colourEn savoir plus