Imprimer
Portable/wall group lockout box, Prinzing
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
046134
BRDY046134EA 160 EUR
BRDY046134
Portable/wall group lockout box, Prinzing
Cadenas de sécurité
This box accommodates 8 to 10 locks (depending on lock size).
  • Made of yellow powder-coated metal (mild sheet steel)
  • Can be used alone or in combination with Industrial Strength lockout station
Informations de commande: Contents not included.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable