Imprimer

Anti-DAD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

BS-6800R-A555
BOSSBS-6800R-A555EA 893 EUR
BOSSBS-6800R-A555
Anti-DAD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
Component of the N-oligosaccharyl transferase enzyme which catalyzes the transfer of a high mannose oligosaccharide from a lipid-linked oligosaccharide donor to an asparagine residue within an Asn-X-Ser/Thr consensus motif in nascent polypeptide chains. N-glycosylation occurs cotranslationally and the complex associates with the Sec61 complex at the channel-forming translocon complex that mediates protein translocation across the endoplasmic reticulum (ER). Loss of the DAD1 protein triggers apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DAD1
Clonality:En savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR