Imprimer

Anti-AHCY Rabbit Polyclonal Antibody (Cy5.5®)

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

BS-5029R-CY5.5
BOSSBS-5029R-CY5.5EA 909 EUR
BOSSBS-5029R-CY5.5
Anti-AHCY Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
Adenosylhomocysteine is a competitive inhibitor of S-adenosyl-L-methionine-dependent methyl transferase reactions; therefore adenosylhomocysteinase may play a key role in the control of methylations via regulation of the intracellular concentration of adenosylhomocysteine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AHCY
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR