Imprimer

Anti-SAHH Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

BS-5029R-A750
BOSSBS-5029R-A750EA 899 EUR
BOSSBS-5029R-A750
Anti-SAHH Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
Adenosylhomocysteine is a competitive inhibitor of S-adenosyl-L-methionine-dependent methyl transferase reactions; therefore adenosylhomocysteinase may play a key role in the control of methylations via regulation of the intracellular concentration of adenosylhomocysteine.

Type: Primary
Antigen: SAHH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 25-125/432
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Others
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR