Imprimer

Anti-CLDN4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

BS-2790R-A488
BOSSBS-2790R-A488EA 901 EUR
BOSSBS-2790R-A488
Anti-CLDN4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact made up of network of strands that act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLDN4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR