Imprimer

Anti-GBA2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

BS-13297R-A350
BOSSBS-13297R-A350EA 893 EUR
BOSSBS-13297R-A350
Anti-GBA2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
Non-lysosomal glucosylceramidase that catalyzes the conversion of glucosylceramide (GlcCer) to free glucose and ceramide. Involved in sphingomyelin generation and prevention of glycolipid accumulation. May also catalyze the hydrolysis of bile acid 3-O-glucosides, however, the relevance of such activity is unclear in vivo. Plays a role in central nevous system development. Required for proper formation of motor neuron axons.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GBA2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
En savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR