Imprimer
Anti-NMUR1 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
BS-11420R-CY7
BOSSBS-11420R-CY7EA 900 EUR
BOSSBS-11420R-CY7
Anti-NMUR1 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
Neuromedin U is a neuropeptide with high activity on smooth muscle. It is widely expressed in gastrointestinal systems and central nervous system (CNS). Peripheral activities of neuromedin U include smooth muscle stimulation, ion transport alterations in the gut and the regulation of local blood flow and adrenocortical function. Neuromedin U receptors 1 and 2 (NMUR1 and NMUR2) are multi-pass membrane proteins that belong to the G-protein coupled receptor 1 family of proteins. Both NMUR1 and NMUR2 act as receptors for the neuromedin U neuropeptide. NMUR1 is detected in peripheral organs,En savoir plus