Imprimer

Anti-EDN1 Rabbit Polyclonal Antibody (Cy3®)

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

BS-0202R-CY3
BOSSBS-0202R-CY3EA 900 EUR
BOSSBS-0202R-CY3
Anti-EDN1 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Endothelins (ET) show potent constrictor activity in vascular and non-vascular smooth muscle. This family of 21-amino acid peptides exists in at least three isoforms - ET-1, ET-2, and ET-3, and is produced in endothelial and epithelial cells. ET's can mediate biological effects in cells and tissues, and have been shown to bind to an ET receptor in the lung, kidney, heart, and liver. Endothelin 1 is expressed in lung, placental stem villi vessels and in cultured placental vascular smooth muscle cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EDN1
Clonality:En savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR