Imprimer
Tiges de filtrage
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
N1505-32 N1520-28 N1510-28 N1505-28
BOHLN1505-32EA 164 EUR
BOHLN1505-32 BOHLN1520-28 BOHLN1510-28 BOHLN1505-28
Tiges de filtrage
Filtres Tubes filtrants
These filtering rods are made of microporous PTFE.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Diameter and length are nominal dimensions and can contain a machining tolerance.