Imprimer
Ring wrenches, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
D701-24
BOHLD701-24EA 15.9 EUR
BOHLD701-24
Ring wrenches, BOLA
Clés à molette
These PP ring wrenches can be used for opening and closing multiple distributors for barrels.
  • Very good chemical resistance
  • Temperature resistant from –20 to +110 °C