Imprimer
Hose connectors, with nut
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
D581-02 D581-04 D582-04 D581-06 D582-02 D581-08
BOHLD581-02EA 10.2 EUR
BOHLD581-02 BOHLD581-04 BOHLD582-04 BOHLD581-06 BOHLD582-02 BOHLD581-08
Hose connectors, with nut
Raccords et connecteurs
GL 14 and GL 18 hose connectors, made of PFA, GL 25 and GL 32 hose connectors, made of PTFE, with elastic sealing lip and FPM O-ring and nut made of PPS.
  • Available as straight or bent type
  • Universal chemical resistance
  • Maximum temperature up to 250 °C