Imprimer
Vessel for pathology dispatch
Fournisseur: BOETTGER PAUL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
08.313.1001 08.323.1001 08.343.1001
BOET08.313.1001EA 190 EUR
BOET08.313.1001 BOET08.323.1001 BOET08.343.1001
Vessel for pathology dispatch
Récipients de collecte d'échantillon Conteneurs à usage universel
Polypropylene, extremely clear with HDPE screw cap.
  • Tight fitting screw cap
  • Stable base
  • 35 ml vessel used to protect the 20 ml vessel, 100 ml vessel used to protect the 50 ml vessel
Specially rounded base to facilitate removal of small tissue samples.