Imprimer
Tube rotator
Fournisseur: BOEKEL SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
260650
BOEI260650EA 1380 EUR
BOEI260650
Tube rotator
Agitateurs Agitateurs rotatifs
Five carousel options with adjustable speed and tilt angles provide greater versatility.
Timer feature offers time down or time up counting. Digital microprocessor controlled. Supplied with a 14×50 ml tube carousel. 2-year warranty.