Imprimer

Anti-CD14 Mouse Monoclonal Antibody (Biotin) [clone: LPSR/927]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

BNCB0927-500 BNCB0927-100
BNCB0927-500EA 560 EUR
BNCB0927-500 BNCB0927-100
Anti-CD14 Mouse Monoclonal Antibody (Biotin) [clone: LPSR/927]
Anticorps
Recognizes a protein of 55 kDa, identified as CD14 (also known lipopolysaccharide receptor). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: LPSR/927
En savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR