Imprimer
Anti-CD3E Mouse Monoclonal Antibody (CF594) [clone: B-B12]
Fournisseur: Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
BNC941052-500 BNC941052-100
BNC941052-500EA 560 EUR
BNC941052-500 BNC941052-100
Anti-CD3E Mouse Monoclonal Antibody (CF594) [clone: B-B12]
Anticorps
Reacts with five invariable CD3 chains (designated as and ) with molecular weight ranging from 16-28 kDa. CD3 is expressed, typically at high levels, on peripheral T cells and majority of T cell neoplasms. Thymocytes express CD3 at different level on the cell surface in the course of differentiation and, in cortical thymus, CD3 is predominantly Intracytoplasmic. The CD3 complex is closely associated at the lymphocyte cell surface with T cell antigen receptor (TCR) and is involved in transducing antigen-recognition signals into cytoplasm of T cells and in regulating the cell surfaceEn savoir plus