Imprimer
Anti-Nucleolar / Nucleoli Mouse Monoclonal Antibody (CF640R) [clone: NM95]
Fournisseur: Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
BNC400095-500 BNC400095-100
BNC400095-500EA 560 EUR
BNC400095-500 BNC400095-100
Anti-Nucleolar / Nucleoli Mouse Monoclonal Antibody (CF640R) [clone: NM95]
Anticorps
This MAb is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. MAb NM95 recognizes an antigen associated with the nucleoli in human cells. It can be used to stain the nucleoli in cell or tissue preparations and can be used as a marker of the nucleoli in subcellular fractions. It produces aEn savoir plus