Imprimer
Cleanroom wipes, CHOICE® 900
Fournisseur: BERKSHIRE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
CH909.10
BERKCH909.10EA 881 EUR
BERKCH909.10
Cleanroom wipes, CHOICE® 900
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
Cleanroom wipes made of 100% continuous filament knit polyester, ultrasonically sealed edge for reduced fiber contamination.
  • Critically low particles, fibers, ions and extractables
  • High abrasion resistant
  • Excellent absorbency
  • Chemically compatible with IPA, Acetone and other solvents
Designed for use in ISO Class 3 and higher environments.
Suitable for breakdown cleans in fab and suite construction, oxidation, metallisation or CVD deposition areas, chamber cleaning and CMP processing, stencil and other print roll cleaning applications, cleaning ofEn savoir plus