Imprimer
Digital refractometer, RFM742-M
Fournisseur: BELLINGHAM STANLEY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
19-20
BELL19-20EA 11430 EUR
BELL19-20
Digital refractometer, RFM742-M
Réfractomètres Réfractomètres de paillasse
The RFM742-M refractometers are robust fully automatic instruments suited to the food, sugar and beverage industries where temperature control is not required, such as measuring sugar (°Brix) concentrations in the food processing industry.
  • Automatic sense 'hands free' measurements
  • Simple audit trail: Date, time and batch number
  • Alpha-numeric keypad for easy data entry
  • Flat sapphire prism surface for easy-cleaning
  • Selectable read type: Continual, single, intelligent auto-sense
May be used in the traditional food, sugar and beverage applications, and inEn savoir plus