Imprimer
Media bottles, graduated, with screw cap
Fournisseur: BELLCO GLASS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5636-00125 5636-00250 5636-00533 5636-38500
BELC5636-00125EA 194 EUR
BELC5636-00125 BELC5636-00250 BELC5636-00533 BELC5636-38500
Media bottles, graduated, with screw cap
Flacons Flacons pour milieux de culture
Manufactured from type 1, class B borosilicate glass.
  • White graduations
  • Marking spot
Supplied with a 33-430 nontoxic, rubber-lined black phenolic cap Large opening and sloping shoulders facilitate pouring and cleaning.