Imprimer
Microcarrier spinner flasks
Fournisseur: BELLCO GLASS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1965-00100 1965-00250 1965-00500 1965-01000 1965-03000 1965-06000 1965-15000 1965-02500 1965-08000
BELC1965-00100EA 548 EUR
BELC1965-00100 BELC1965-00250 BELC1965-00500 BELC1965-01000 BELC1965-03000 BELC1965-06000 BELC1965-15000 BELC1965-02500 BELC1965-08000
Microcarrier spinner flasks
Ballons Flacons réacteurs
Spinner flasks promotes the optimal growth of suspension and microcarrier culture.
  • Impeller height adjusts through compression fitting
  • Indented bottom (up to 1 L) prevents accumulation of microcarrier beads under impeller