Imprimer
Serological pipettes, Shortie
Fournisseur: BELLCO GLASS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1224-05010
BELC1224-05010EA 541 EUR
BELC1224-05010
Serological pipettes, Shortie
Pipettes Pipettes sérologiques
These serological pipettes are manufactured from type 1 borosilicate glass.
  • Shorter length facilitates use under biological hoods
  • Plain top promotes rapid, even flow (use rubber bulb to fill)
  • Calibrated to tip
  • Tempered tip; to deliver, blow-out
  • Will withstand repeated autoclaving