Imprimer
Autoclave deodoriser, triple strength, Odo-Clave®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
H131990000
BELAH131990000EA 595 EUR
BELAH131990000
Autoclave deodoriser, triple strength, Odo-Clave®, SP Bel-Art
Déodorants et désodorisants
This heat-released deodorant pads neutralise autoclave odors with a pleasant scent.
  • Simply open foil pouch and place pad in bag prior to autoclaving
  • Pads have a three year life span; will last longer if stored in a sealed container
  • Triple strength clove releases three times the deodorant of the standard pads while costing less than two standard pads
W×L: 57×57 mm
Cdt: Packed 10 pads per bag, 10 bags per box.