Imprimer
Desiccator cabinets, auto, horizontal, secador®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
F420740220
BELAF420740220EA 3820 EUR
BELAF420740220
Desiccator cabinets, auto, horizontal, secador®, SP Bel-Art
Dessiccateurs Armoires de dessiccation
Secador® 4.0 auto desiccator cabinets are made from moulded Durastar® co-polyester which blocks 99% of UV light and resists staining, crazing and chemical attack.
  • Large, pull down, oven style door with easy-to-view digital hygrometer
  • Two perforated shelves facilitate circulation of dry air
  • Colour-indicating silica gel allows quick visual confirmation of desiccant status
Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with Secador® Auto-Desiccator cabinets. Automatic dehumidification andEn savoir plus