Imprimer
Fritware immersion filters, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
F136700000
BELAF136700000EA 162 EUR
BELAF136700000
Fritware immersion filters, SP Bel-Art
Filtres Tubes filtrants
Immersion filters for filtration involving recovery of solids without having to transfer material to funnels.
  • High density PE plastic coarse porosity disc and body
  • Use directly or precoat for fine filtration
  • Has a serrated tubing connector