Imprimer
BD Eclipse™ Needles
Fournisseur: BD Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
305781 305892 305770 305771 305895 305784 305760 305891 305894
BDAM305781EA 71.6 EUR
BDAM305781 BDAM305892 BDAM305770 BDAM305771 BDAM305895 BDAM305784 BDAM305760 BDAM305891 BDAM305894
BD Eclipse™ Needles
Aiguilles pour seringues
The BD Eclipse™ SmartSlip™ is a single use sterile needle with a safety mechanism that covers the needle point after use. In the activated position, the needle cover guards against accidental needlestick during normal handling and disposal of the used needle/syringe.
  • The SmartSlip™ Technology is specially designed for safe assembling of BD Eclipse™ needles to Luer slip syringes
  • Bevel oriented to the safety cover to allow low angle injections
  • By assembling BD Eclipse™ needles to a luer slip syringe, an audible click indicates a safely assembled needle
En savoir plus