Imprimer
Aiguilles de remplissage universelles
Fournisseur: BD Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
305211 305181 303129 303129UK
BDAM305211EA 201 EUR
BDAM305211 BDAM305181 BDAM303129 BDAM303129UK
Aiguilles de remplissage universelles
Aiguilles pour seringues
BD blunt fill and blunt filter needles are specifically designed to reduce the risk of needlestick injuries during medication preparation.
  • The blunt fill needle is designed for vial access and available with a BD Luer-Lok™ connection
  • The blunt filter is designed for safe ampoule medication preparation
  • BD blunt fill needles have female luer fittings, which mate to male luer fittings
  • BD blunt filter needle has a 5-micron filter for particles such as glass and plastic fragments, for safer medication delivery
  • Natural rubber latex is not part of the material
En savoir plus