Imprimer
BD PrecisionGlide™ Needles
Fournisseur: BD Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
305196
BDAM305196EA 527 EUR
BDAM305196
BD PrecisionGlide™ Needles
Aiguilles pour seringues
BD PrecisionGlide™ Needle technology incorporates a thin wall, tri-bevel tip and a proprietary lubricant which is designed to provide ease of penetration.
  • Using a BD thin-wall needle may enable you to use a smaller gauge needle as the flow rate is similar to a BD regular-wall needle one gauge larger
  • BD has 20 years experience manufacturing thin-wall needles and meet ISO standards for cannula stiffness and resistance to breakage
  • 69% of patients said they would prefer to receive a skin injection with a smaller-diameter needle