Imprimer

Hotplate

Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

RC2240Q
BARNRC2240QEA 2240 EUR
BARNRC2240Q
Hotplate
Appareils chauffants Plaques chauffantes
External-controlled hotplates with porcelain-coated stainless-steel top.

  • Isolation of controls allows operation in hazardous environments
  • Large top plate surfaces accommodate up to 40 lb. (18.1 kg) loads
  • Power light indicates when control is operating
  • Large capacity with excellent temperature stability
  • Percentage input controller delivers infinite temperature selection
  • Porcelain-coated, stainless-steel top offers chemical and stain resistance
  • Chemical-resistant cover protects line cord
  • Stainless-steel case offers maximum resistance to corrosion

TheseEn savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR