Imprimer
Safety wash bottles, wide neck, venting, multilingual
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
WGW537VTMLA WGW538VTMLA WGW539VTMLA WGW541VTMLA WGW601PMLA
AZLOWGW537VTMLAEA 55.7 EUR
AZLOWGW537VTMLA AZLOWGW538VTMLA AZLOWGW539VTMLA AZLOWGW541VTMLA AZLOWGW601PMLA
Safety wash bottles, wide neck, venting, multilingual
Flacons Pissettes
LDPE wash bottles gives accurate dispensation of liquid.
  • The patented DripLok® vapour venting valve prevents solvent drips
  • Indelibly printed in three languages (English, French and Spanish)
  • Closure feature precision jet-tip aperture for fine control
Wide neck for easy, safe filling. Fitted with colour coded one-piece PP closure that maintain their shape in use.
Certifications: Wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.