Imprimer
Clapets anti-retour, Azlon®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
NWH902P NWH906P
AZLONWH902PEA 62.2 EUR
AZLONWH902P AZLONWH906P
Clapets anti-retour, Azlon®
Vannes Clapets anti-retour
Designed for use in vacuum lines and fluid handling systems.
  • Useful for preventing back pressure in filter flasks
  • Printed arrow indicates the direction of flow
  • Rigid with stepped tapered arms to give firm grip on tubing