Imprimer
Pre-sterilized individually wrapped blue pestle, Axygen®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
PES-15-B-SI
AXYGPES-15-B-SIEA 490 EUR
AXYGPES-15-B-SI
Pre-sterilized individually wrapped blue pestle, Axygen®
Mortiers et pilons Pilons
Durable and inert polypropylene construction
  • Designed to fit 1.5 and 2.0 ml centrifuge tubes
  • Individually wrapped and sterilized