Imprimer
Lloyd and McCown's woody plant medium with vitamins
Marques : Apollo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
PML449B-10L
APOSPML449B-10LEA 68 EUR
APOSPML449B-10L
Lloyd and McCown's woody plant medium with vitamins
Milieux de culture cellulaire
Contains the WPM macro- and micronutrients and vitamins as described by Lloyd and McCown (1981). Plant tissue culture tested.