Imprimer

Cycloheximide 100 mM

Marques : G Biosciences
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,

API-03
API-03EA 273 EUR
API-03
Cycloheximide 100 mM
Cycloheximide

Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).


  • Supplied as 1ml 100mM solution.


G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.


Cycloheximide is an inhibitor of protein synthesis by interfering with the translocation step (movement of two tRNA molecules and mRNA on ribosome) in protein synthesis, thus blocking translational elongation.

Formule: C₁₅H₂₃NO₄
Poids moléculaire: 281.35 g/mol
Numéro MDL: MFCD00082346
Numéro CAS: 66-81-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2aa4423dbded4a11a8b76daa453019131515b275&vid=fe0bbb3aaa025bedcd2b60bfb62c01be3fd77bdd&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR