Imprimer
Fragment Analyzer System, 5400
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
M5312AA
AGLSM5312AAEA 96550 EUR
AGLSM5312AA
Fragment Analyzer System, 5400
Systèmes d'électrophorèse Capillaires pour électrophorèse
Parallel capillary electrophoresis system that integrates with robotic arms for high-throughput labs.
  • Robotic integration - a custom API and powered drawers offer seamless integration with most laboratory automation systems
  • High throughput separations - unattended analysis of more than 2,400 samples every 24 hours
  • High separation resolution - resolve a wide range of DNA and RNA fragments. Achieve as low as 3 bp resolution with high sizing accuracy for fragments under 300 bp
  • Low cost per sample - choose qualitative or quantitative gel kits that best suit
En savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable