Imprimer
Qualitative DNA Kits
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
DNF-905-K0500 DNF-905-K1000 DNF-910-K0500 DNF-910-K1000 DNF-915-K0500 DNF-915-K1000 DNF-920-K0500 DNF-930-K0500 DNF-930-K1000 DNF-935-K0500 DNF-935-K1000
AGLSDNF-905-K0500EA 479 EUR
AGLSDNF-905-K0500 AGLSDNF-905-K1000 AGLSDNF-910-K0500 AGLSDNF-910-K1000 AGLSDNF-915-K0500 AGLSDNF-915-K1000 AGLSDNF-920-K0500 AGLSDNF-930-K0500 AGLSDNF-930-K1000 AGLSDNF-935-K0500 AGLSDNF-935-K1000
Qualitative DNA Kits
Systèmes d'électrophorèse Capillaires pour électrophorèse
DNA fragment analysis for PCR, genotyping, and microsatellite applications.
  • Broad concentration range - all kits have a concentration range of 0.5 to 50 ng/µl
  • Reliable sizing - six different kits to choose from with a sizing range varying from 35 to 20,000 bp
  • Low sample volume - only 2 µl of sample is required, minimising precious sample loss to QC steps
  • Quick separations - electrophoresis times as short as 15 minutes
  • High resolution - separate fragments under 300 bp differing by only 3 bp
The qualitative kits provide automated and accurate sizingEn savoir plus