Imprimer
Genomic DNA Kits
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
DNF-467-0500 DNF-468-0500 DNF-488-0500 DNF-488-1000
AGLSDNF-467-0500EA 1480 EUR
AGLSDNF-467-0500 AGLSDNF-468-0500 AGLSDNF-488-0500 AGLSDNF-488-1000
Genomic DNA Kits
Systèmes d'électrophorèse Capillaires pour électrophorèse
Genomic DNA quality control of whole genome libraries, including sizing of gDNA extractions.
  • Broad concentration range - The HS gDNA 50 kb kit has a concentration range of 0,3 to 12 ng/µl, while the gDNA 50 kb kit covers 25 to 250 ng/µl
  • High and standard sensitivity kits - Minimise sample dilutions and choose the kit that has the right input concentration range for your needs
  • Reliable Sizing - Sizing range from 75 bp to 60 kb ensures accurate and precise sizing of gDNA samples
  • Low sample volume - Only 1 to 2 µl of sample is required, minimising sample loss
En savoir plus