Imprimer
Cloned Pfu DNA polymerase alternative detergents
Marques : AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
600355 600357
AGLS600355EA 474 EUR
AGLS600355 AGLS600357
Cloned Pfu DNA polymerase alternative detergents
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
Cloned Pfu DNA Polymerase for high fidelity and difficult, GC-Rich PCR.
  • Highly thermostable Pfu DNA polymerase possessing 3' to 5' exonuclease proofreading activity that enables the polymerase to correct nucleotide-misincorporation errors (1 error per 770000 bp)
  • Reformulated buffer system for increased economy with the same high performance as the original Pfu DNA polymerase
  • Blunt ends
Using Cloned Pfu DNA polymerase in PCR experiments results in blunt-ended products, which are ideal for cloning into blunt-ended vectors. Cloned Pfu DNA Polymerase, derived from the hyperthermophilic archaea Pyrococcus furiosus exhibits superior thermostability and proofreading properties.
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Application
Learn more