Imprimer
DNA ScreenTape Analysis (ScreenTape and Reagents)
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5067-5582 5067-5583 5067-5586 5067-5588 5067-5589 5067-5590 5067-5602
AGLS5067-5582EA 223 EUR
AGLS5067-5582 AGLS5067-5583 AGLS5067-5586 AGLS5067-5588 AGLS5067-5589 AGLS5067-5590 AGLS5067-5602
DNA ScreenTape Analysis (ScreenTape and Reagents)
Systèmes d'électrophorèse Capillaires pour électrophorèse
DNA fragment or library analysis for sample quality control of samples up to 5000 bp.
  • A broad size range allows analysis of DNA fragments and libraries from 35 to 5000 base pairs
  • Fast results are obtained, with data delivered in approximately 1 minute per sample
  • Minimal sample consumption is required as only 1 µl of DNA sample is needed
  • DNA sample detection is possible down to 0,1 ng/µl
  • Ready-to-use ScreenTape devices, pre-packaged reagents, and automated data analysis offer an easy to use system with reproducible DNA sizing and quantification results
  • No
En savoir plus